Learn More
Cole-Parmer™ Replacement Cylinder for Large Grinding Vial Sets
Alternative stainless steel center cylinder for 6801 Large Grinding Vial Set.
Brand:  Cole-Parmer™ 6802
Product Code. 15300482
  • 7070.00 DKK / Each
Estimated Shipment: 25-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • Replaces polycarbonate center cylinders when sample contact with plastic is not acceptable.
  • Made of nonmagnetic 440C stainless steel.
  • Withstands liquid nitrogen temperatures.
  • Autoclavable and reusable.
Specifications
Specifications
Center Cylinder
Reusable
6801 Large Grinding Vial Set (61043-10)
1 Ea.
Autoclavable
Replaces Polycarbonate Cylinder When Sample Contact with Plastic Cannot be Used
Stainless Steel (440 C)