Learn More
Cole-Parmer™ Replacement Cylinders for Mid-Size Grinding Vial Sets
Replacement Cylinders for 6881 mid-size grinding vial set, 6883 mid-size low-chrome grinding vial set, and 6885 Mid-size poly-vial set.
Brand: Cole-Parmer™ 6881C4
This item is not returnable.
View return policy
Description
- Withstands liquid nitrogen temperatures.
- Are disposable or may be cleaned with hot water and detergent or RNase/DNase-free solution.
- Transparent plastic center cylinder allows visual checking of the grinding progress.
Specifications
Center Cylinder | |
Mid Size, Cr-Free, Poly-Vial Grinding Vial Set | |
Polycarbonate | |
4 Pk. |