Learn More
Bollé Safety Slam Clear Safety Glasses
Stylish and easy-to-wear. The SLAM model provides protection at all timeswhile offering unbeatable comfort
Brand:  Bollé Safety SLAPSI
369.57 DKK valid until 2025-07-04
Use promo code "24152" to get your promotional price.
Product Code. 11913584
  • 488.00 DKK / Pack of 10
Gear Up for Safety
Use promo code 24131 at checkout
View Available Promotions
Only null left
Estimated Shipment: 04-07-2025
Description
  • Wrap-around fit
  • All-round vision
  • Comfortable straight temples
  • Adjustable cord
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Anti-fog, Scratch-resistant
Safety Spectacles
10/Pk.
Documents
Specification Sheets
Certificates
Promotions
Promotions
  • Multiple Manufacturers
    Save up to 60% on gloves, apparel, and more
    Promo Code: 24131
    Expires2025-06-08
  • Multiple Manufacturers
    Your vision deserves the best protection.
    Promo Code: 24152
    Expires2025-07-04
    New Offer
    Shop Now
Product Content Correction
Product Title
Bollé Safety Slam Clear Safety Glasses > Lens Coating: Anti-Fog & Anti-Scratch Coating