Learn More
Lenz Laborglasinstrumente™ Spiral Springs
Brand:  Lenz Laborglasinstrumente™ 01102825
 View more versions of this product
Product Code. 10221071
  • 238.00 DKK / Pack of 10
Estimated Shipment: 28-05-2025
to see stock.
Explore more special offers
Specifications
25 mm
Documents
One moment while we fetch your results.
Product Title
Lenz Laborglasinstrumente™ Spiral Springs > Length: 25mm