Learn More
Spironolactone, MedChemExpress
MedChemExpress Spironolactone (SC9420) is an orally active aldosterone mineralocorticoid receptor antagonist with an IC50 of 24 nM. Spironolactone is also a potent antagonist of androgen receptor with an IC50 of 77 nM. Spironolactone promotes autophagy in podocytes.
Brand: Medchem Express HY-B0561
Description
Spironolactone (SC9420) is an orally active aldosterone mineralocorticoid receptor antagonist with an IC50 of 24 nM. Spironolactone is also a potent antagonist of androgen receptor with an IC50 of 77 nM. Spironolactone promotes autophagy in podocytes.Specifications
Spironolactone | |
White | |
C24H32O4S | |
SC9420 | |
C[C@@]12[C@](OC3=O)(CC3)CC[C@@]1([H])[C@@]([C@@H](CC4=CC5=O)SC(C)=O)([H])[C@]([C@]4(CC5)C)([H])CC2 | |
416.57 | |
Research | |
Cancer-programmed cell death |
52-01-7 | |
Research | |
1 g | |
DMSO : ≥ 50 mg/mL (120.03 mM) ∣H2O : 0.1 mg/mL (0.24 mM; Need ultrasonic) | |
416.57 | |
99.58% | |
Solid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Research purposes only