accessibility menu, dialog, popup
Test
Search
Order Status
Quick Order
Support
Sign In
chevron-down
chevron-down
Learn More
Home
Products
Mixers
Mortars and Pestles
Haldenwanger™ Spouted Mortar
12005133
Print
Haldenwanger™ Spouted Mortar
Diameter (Metric) Outer: 50mm
Brand:
Haldenwanger™ 55/00
View more versions of this product
Product Code.
12005133
123.00 DKK
/ Each
Quantity
Estimated Shipment: 23-12-2024
Log in
to see stock.
Explore more special offers
Add to basket
This item is not returnable.
View return policy
Specifications
Capacity (Metric)
20 mL
Product Type
Mortar
Type
Mortar
Diameter (Metric) Outer
50mm
Includes
With spout