Learn More
Haldenwanger™ Squat Form Porcelain Crucibles Without Lid
Use these dishes for a variety of applications. Haldenwanger™ Squat Form Porcelain Crucibles feature a smooth internal finish, constructed from optimized laboratory porcelain.
Brand:  Haldenwanger™ L 79/2A
 View more versions of this product
Product Code. 12346487
  • 1095.00 DKK / Pack of 10
Estimated Shipment: 11-06-2025
Explore more special offers
Description
  • Available in a variety of sizes
  • Sizes up to 175mL glazed except rim
  • 1,000mL and larger are glazed except outside base
  • Complies with DIN VDE 0335, type C 100
Specifications
Specifications
Porcelain
50 mL
35mm
Glazed
60mm
Squat Form Porcelain Crucibles
Product Title
Haldenwanger™ Squat Form Porcelain Crucibles Without Lid > Capacity: 50mL