Learn More
Tocris Bioscience™ SR 3576
Highly potent and selective JNK3 inhibitor
Brand:  Tocris Bioscience™ 5044/50
4310.04 DKK valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18747048
  • 4855.00 DKK / 50mg
Explore more special offers
This item is not returnable. View return policy
Description
Description
SR 3576 is a highly potent and selective JNK3 inhibitor (IC50 = 7 nM); exhibits > 2800-fold selectivity over p38.
Specifications
Specifications
>97%
SR 3576
SR 3576
50 mg
501.53
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
For Research Use Only