Learn More
Tocris Bioscience™ SR 3576
Highly potent and selective JNK3 inhibitor
Brand:  Tocris Bioscience™ 5044/10
1118.76 DKK valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18761779
  • 1265.00 DKK / 10mg
Explore more special offers
Description
Description
SR 3576 is a highly potent and selective JNK3 inhibitor (IC50 = 7 nM); exhibits > 2800-fold selectivity over p38.
Specifications
Specifications
>97%
SR 3576
SR 3576
10 mg
501.53
Special Offers
For Research Use Only