missing translation for 'onlineSavingsMsg'
Learn More

SREBP2 Antibody (SREBP2/1580), Janelia Fluor™ 646, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP3-14186JF646

Product Code. 18310836

  • 4437.25 DKK / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

SREBP2 Monoclonal antibody specifically detects SREBP2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications

Specifications

SREBP2
Monoclonal
Janelia Fluor 646
50mM Sodium Borate
Mouse
Protein A or G purified
RUO
Primary
Human
Antibody
IgG1 κ
Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
SREBP2/1580
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
BHLHD2, bHLHd2sterol regulatory element-binding protein 2, Class D basic helix-loop-helix protein 2, SREBP2, SREBP2SREBP-2, sterol regulatory element binding transcription factor 2, Sterol regulatory element-binding transcription factor 2
Recombinant full-length human SREBP2protein (Uniprot: Q12772)
0.1 mL
metabolism
6721
Store at 4°C in the dark.
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers