Learn More
BINDER™ Stacking Frame
Stable, vibration-free stacking frame on casters with stop brake for safe stacking of two CB series units.
Brand: BINDER™ 9051-0041
This item is not returnable.
View return policy
Description
- For safe stacking of two CB series units
- Suitable for:
- CB-S 260
- CB 260
Specifications
Stacking frame - stable, vibration-free stacking frame on casters with stop brake for safe stacking of two CB series units | |
Steel | |
For stability, vibration-free stacking | |
841 x 930 x 1239 mm |
BINDER Chambers CB260, CB-S260, CBF 260 | |
Stacking Frame | |
Casters |
Product Content Correction
Product Title
BINDER™ Stacking Frame > For Use With: BINDER units CB260 and CB-S260
Spot an opportunity for improvement?Share a Content Correction