Learn More
Memmert™ Stacking Set for Stacking of Two Units of Same Size
Stacking Set for stacking of appliances of same size with subframe
Brand:  Memmert™ ICO240B48129
 View more versions of this product
Product Code. 17139984
  • 4420.00 DKK / Each
Only null left
Estimated Shipment: 19-08-2025
Add to basket
This item is not returnable. View return policy
Description
Description
  • Only in connection with subframe B33509 or B43598
  • Consisting of stacking corners
  • One connecting plate for the rear, two wall brackets
Specifications
Specifications
ICO240
Two Units of Same Size
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert™ Stacking Set for Stacking of Two Units of Same Size > ICO240