Learn More
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | ||||||
10303611
|
Bochem™
1150 |
115 mm |
32.50 DKK
Each |
Estimated Shipment: 10-06-2025
Log in to see stock. |
||||||
10484101
|
Bochem™
1151 |
130 mm |
33.75 DKK
Each |
Estimated Shipment: 02-06-2025
Log in to see stock. |
||||||
10663911
|
Bochem™
1152 |
145 mm |
36.45 DKK
Each |
Estimated Shipment: 21-07-2025
Log in to see stock. |
||||||
Description
- Manufactured from stainless steel
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Bochem™ Stainless Steel 1:2 Tissue Forceps