Learn More
VACUUBRAND™ Stainless Steel Butterfly Passage Valve
Compact, economic design, easy to grip, lever-operated with position indicator
Brand: VACUUBRAND™ 20665005
3348.75 DKK valid until 2025-04-13
Use promo code "24122" to get your promotional price.
This item is not returnable.
View return policy
Description
The VS und VS C series valves are butterfly-type valves. A circular valve plate with a sealing ring on the circumference rotates around an axis at right angle to the valve axis. As a result, the valves leave virtually the entire cross section free, offering very good conductance. The valve bodies are made of one piece, machined from solid material, and require (in addition to the valve head seal) only one elastic shaft seal for rotational movement of the shaft. The valves provide low leakage and minimal degassing rate. The VS C series features a valve seal made of perfluorelastomer material (ffKM).
Specifications
KF VS25 / DN | |
Stainless Steel | |
1 Each | |
Butterfly Passage Valve |
Special Offers
-
Multiple Manufacturers
Precision at Your Fingertips
Master liquid handling with ease.Promo Code: 24122Expires: 2025-04-13
Days
10Hours
3Minutes
7Seconds
52
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
VACUUBRAND™ Stainless Steel Butterfly Passage Valve > For use with: KF VS25 / DN
Spot an opportunity for improvement?Share a Content Correction