Learn More
Nickel Electro™ Stainless Steel Flask Clip
For personalised clip arrangement, clips are supplied with all necessary fixings to enable them to be attached to the Perforated Tray. Nickel Electro™ Stainless Steel Flask Clip is suitable for flasks and can be autoclaved.
Brand:  Nickel Electro™ FC0025
109.06 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 11736577
  • 131.00 DKK / Each
Only null left
Estimated Shipment: 03-10-2025
Description
  • Highly flexible elastic design allowing glassware to easily slide into the clip and yet remain secure during mixing
  • No sharp edges to cut or score glassware
  • Can be autoclaved
  • Plastic base
  • Easy to assemble and remove Flask Clips from Perforated tray
Specifications
Specifications
25 mL Conical Flask
Flask Clip
Stainless Steel
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Nickel Electro™ Stainless Steel Flask Clip > For Use With: 25mL Conical Flask