Learn More
KGW Isotherm™ Stainless Steel Dewar Flasks
Brand:  KGW Isotherm™ 2303
 View more versions of this product
Product Code. 10421811
  • 3835.00 DKK / Each
Estimated Shipment: 19-05-2025
Explore more special offers
Specifications
312mm
2 L
122 mm
Stainless Steel
Certificates
View accessory products
Product Title
KGW Isotherm™ Stainless Steel Dewar Flasks > Capacity: 2L