Learn More
KGW Isotherm™ Stainless Steel Dewar Flasks
Brand:  KGW Isotherm™ 2302
 View more versions of this product
Product Code. 10662741
  • 5055.00 DKK / Each
Estimated Shipment: 07-04-2025
to see stock.
Explore more special offers
Specifications
190mm
3 L
200 mm
Stainless Steel
Documents
One moment while we fetch your results.
Certificates
View accessory products
Product Title
KGW Isotherm™ Stainless Steel Dewar Flasks > Capacity: 3L