Learn More
Socorex™ Stainless Steel Injection Needles
Brand:  Socorex™ 371.16100
Product Code. 11877283
  • 574.00 DKK / Pack of 12
Explore more special offers
Specifications
Autoclavable
Stainless Steel
100 mm
Injection Needle
16 G
1.6 mm
Product Title
Socorex™ Stainless Steel Injection Needles > Gauge: 16G; Length: 100mm