Learn More
Bochem™ Stainless Steel Laboratory Scissors
Manufactured from stainless steel
59.75 DKK - 93.55 DKK
Specifications
Product Type | Laboratory Scissors |
---|---|
Material | Stainless Steel |
Tip Style | Sharp/Round |
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | ||||||
10633652
|
Bochem™
4000 |
100 mm |
59.75 DKK
Each |
Estimated Shipment: 15-08-2025
Log in to see stock. |
||||||
10305961
|
Bochem™
4001 |
115 mm |
65.10 DKK
Each |
Estimated Shipment: 06-08-2025
Log in to see stock. |
||||||
10214611
|
Bochem™
4005 |
165 mm |
86.05 DKK
Each |
Estimated Shipment: 18-08-2025
Log in to see stock. |
||||||
10223941
|
Bochem™
4006 |
180 mm |
93.55 DKK
Each |
Estimated Shipment: 15-08-2025
Log in to see stock. |
||||||
10490923
|
Bochem™
4002 |
130 mm |
76.70 DKK
Each |
Estimated Shipment: 29-07-2025
Log in to see stock. |
||||||
10326101
|
Bochem™
4003 |
145 mm |
73.35 DKK
Each |
Estimated Shipment: 21-08-2025
Log in to see stock. |
||||||
10491113
|
Bochem™
4004 |
150 mm |
80.70 DKK
Each |
Estimated Shipment: 01-09-2025
Log in to see stock. |
||||||
Description
- Three types: round-round, sharp-round, and sharp-sharp
- Stainless steel
Specifications
Laboratory Scissors | |
Stainless Steel | |
Sharp/Round |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Bochem™ Stainless Steel Laboratory Scissors