Learn More
SPEX CertiPrep™ Stainless Steel Long Cryogenic Transfer Hose
Useful for transferring cryogenic fluids such as liquid nitrogen.
Brand: SPEX CertiPrep™ 6907
This item is not returnable.
View return policy
Description
- Flexible stainless steel construction.
- Withstands liquid nitrogen temperatures.
- Fitted with a 3/8 in. (9.5 mm) NPT male fitting on one end and aCGA295 female fitting on the other end.
- Compatible with 6905 Portable Cryogenic Dewar and 6820 Auto-Fill System.
Specifications
6905 Portable Cryogenic Dewar and 6820 Auto-Fill System. | |
6 ft. | |
1.8 m |