Learn More
Fisherbrand™ Stainless Steel Manual Crimper for clean room, adjustable
Provides a reproducible, secure vial closure
7960.00 DKK - 9940.00 DKK
Product Code | Brand | Diameter (Metric) Cap | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Cap | Price | Quantity & Availability | ||||||
11737746
|
Fisherbrand™
11 06 2225 |
11 mm |
8415.00 DKK
Each |
Estimated Shipment: 12-08-2025
Log in to see stock. |
||||||
11757746
|
Fisherbrand™
20 06 2229 |
20 mm |
7960.00 DKK
Each |
Estimated Shipment: 14-08-2025
Log in to see stock. |
||||||
11747746
|
Fisherbrand™
13 06 2227 |
13 mm |
7960.00 DKK
Each |
Estimated Shipment: 12-08-2025
Log in to see stock. |
||||||
12686835
|
Fisherbrand™
32 06 2231 |
32 mm | N/A | |||||||
Description
- Stainless Steel
- Sterilizable for cleanroom applications
- Manual
- Autoclavable
Specifications
Stainless Steel |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Fisherbrand™ Stainless Steel Manual Crimper for clean room, adjustable