Learn More
Welch Ilmvac™ Stainless Steel Metal Vacuum Hose
Brand:  Welch Ilmvac™ 710765
1083.33 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 10497281
  • 1115.00 DKK / Each
Only null left
Estimated Shipment: 22-09-2025
Specifications
DN KF IN and EX Connectors
100 cm
Metal Vacuum Hose
Stainless Steel
DN 25
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Welch Ilmvac™ Stainless Steel Metal Vacuum Hose > Length: 1000mm