Learn More
Sartorius™ Stainless Steel OIML Class E1 Reference Line Weight With DAkkS Calibration Certificate
Brand:  Sartorius™ YCW521-02
5879.24 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10012251
  • 6030.00 DKK / Each
Only null left
Estimated Shipment: 15-07-2025
Specifications
Wood
200 g
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Sartorius™ Stainless Steel OIML Class E1 Reference Line Weight With DAkkS Calibration Certificate > Mass: 200g