Learn More
Sartorius™ Stainless Steel OIML Class F1 Reference Line Weight
Stainless Steel OIML Class F1 Reference Line Weight without DAkkS Calibration Certificate.
Brand:  Sartorius™ YCW323-00
620.75 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10072081
  • 641.00 DKK / Each
Only null left
Estimated Shipment: 29-07-2025
Description
Stainless Steel OIML Class F1 Reference Line Weight without DAkkS Calibration Certificate.
  • Without DAkkS Calibration Certificate
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Stainless Steel
Plastic
2 g
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Sartorius™ Stainless Steel OIML Class F1 Reference Line Weight > Plastic; Mass: 2 g