Learn More
Sartorius™ Stainless Steel OIML Class F1 Reference Line Weight
Stainless Steel OIML Class F1 Reference Line Weight without DAkkS Calibration Certificate.
Brand:  Sartorius™ YCW553-00
2350.20 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10154031
  • 2415.00 DKK / Each
Only null left
Estimated Shipment: 16-06-2025
Description
Stainless Steel OIML Class F1 Reference Line Weight without DAkkS Calibration Certificate.
  • Without DAkkS Calibration Certificate
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Stainless Steel
Wood
500 g
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Sartorius™ Stainless Steel OIML Class F1 Reference Line Weight > Wood; Mass: 500 g