Learn More
Usbeck Laborgeraete™ Stainless Steel Powder Patel
Brand: Usbeck Laborgeraete™ 3283
This item is not returnable.
View return policy
Specifications
1 Pk. | |
Powder Patel | |
Stainless Steel | |
185 mm |
Product Content Correction
Product Title
Usbeck Laborgeraete™ Stainless Steel Powder Patel > Length: 185mm
Spot an opportunity for improvement?Share a Content Correction