Learn More
Schneider Gerd™ Stainless Steel Right Analysis Spoon Spatula
Use this spatula for a variety of general laboratory applications. Schneider Gerd™ Stainless Steel Right Analysis Spoon Spatulas are ideal for use with a wide range of compounds.
34.00 DKK - 80.00 DKK
Specifications
Quantity | 1 Ea. |
---|---|
Product Type | Right Analysis Spoon Spatula |
Blade Material | Steel |
Material | Stainless Steel |
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
10316644
|
Schneider Gerd™
212 |
150 mm |
34.00 DKK
Each |
In Stock
Log in to see stock. ![]() |
|||||
10294624
|
Schneider Gerd™
213 |
180 mm |
48.45 DKK
Each |
In Stock
Log in to see stock. ![]() |
|||||
10001414
|
Schneider Gerd™
214 |
210 mm |
53.60 DKK
Each |
Estimated Shipment: 23-09-2024
Log in to see stock. |
|||||
10735735
|
Schneider Gerd™
215 |
250 mm |
80.00 DKK
Each |
In Stock
Log in to see stock. ![]() |
|||||
Description
- Spoons collect and transport dry substances or liquids
- Spatulas are ideal for working with powders, stirring and mixing
- Perfect for miniscule amounts of chemicals
- Two end shapes, offer increased versatility
- Polished stainless steel implements are effortlessly cleaned and corrosion resistant
Specifications
1 Ea. | |
Right Analysis Spoon Spatula | |
Steel | |
Stainless Steel |