Learn More
Swann-Morton™ Stainless Steel Scalpel Blades, 100-pack
Operate using Swann Morton™ Stainless Steel Scalpel Blades to ensure precision;
Brand:  Swann-Morton™ 0305
 View more versions of this product
Product Code. 12991635
  • 241.00 DKK / Pack of 100
Only null left
Estimated Shipment: 25-08-2025
Description
  • Sterile
  • Stainless Steel
Specifications
Specifications
Stainless Steel
# 15
The # 15 Blade fit Handles # 3, # 3L, # 3 Graduated, # 5B, # 7, # 9, # B3 and # B3L
10/Pk.
Scalpel Blade
15 mm
BS EN 7740
Certificates
View accessory products
Promotions
Promotions
Product Content Correction
Product Title
Swann-Morton™ Stainless Steel Scalpel Blades, 100-pack > Fits Model: No. 15; Length: 15mm