Learn More
Memmert™ Stainless Steel Slide-In Bottom Drip Tray for SF110, 15 mm
Non-perforated slide-in drip trays constructed of stainless steel.
Brand:  Memmert™ SF110B02073
Product Code. 17528085
  • 1020.00 DKK / Each
Estimated Shipment: 27-06-2025
Explore more special offers
Description
  • Accessory
  • May affect temperature distribution
  • Constructed of stainless steel
  • Rim height: 15 mm
Specifications
Specifications
SF110 Sterilizer
Drip Tray
Rim: 15 mm
Stainless Steel
Documents
One moment while we fetch your results.
Product Title
Memmert™ Stainless Steel Slide-In Bottom Drip Tray for SF110, 15 mm > For Use With: SF110