Learn More
Memmert™ Stainless Steel Slide-In Bottom Drip Tray for SF55plus, 15 mm
Non-perforated slide-in drip trays constructed of stainless steel.
Brand:  Memmert™ SF55plusB02072
Product Code. 17568335
  • 646.00 DKK / Each
Estimated Shipment: 14-05-2025
to see stock.
Explore more special offers
Description
  • Accessory
  • May affect temperature distribution
  • Constructed of stainless steel
  • Rim height: 15 mm
Specifications
Specifications
SF55plus Sterilizer
Drip Tray
Rim: 15 mm
Stainless Steel
Product Title
Memmert™ Stainless Steel Slide-In Bottom Drip Tray for SF55plus, 15 mm > For Use With: SF55plus