Learn More
Grant Instruments™ Stainless Steel Sloping Lids
Recommended for use above 60°C. Stainless Steel Sloping Lid avoids contamination, reduces evaporation and saves energy.
Brand:  Grant Instruments™ LU6
1693.64 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 11416427
  • 1740.00 DKK / Each
Only null left
Estimated Shipment: 14-10-2025
Description
  • For use with Sub Aqua Pro Series Unstirred Water Bath
  • Triangular-shaped stainless steel with plastic handle
  • Choose from the following available sizes:
    • 5L (LU6)
    • 12L (LU14)
    • 18 or 26L (LU28)
    • 34L (LU36)
    TRUSTED_SUSTAINABILITY
Specifications
Specifications
SAP5, ST5
Stainless Steel
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Grant Instruments™ Stainless Steel Sloping Lids > For Use With: SAP5