Learn More
Grant Instruments™ Stainless Steel Sloping Lids
Recommended for use above 60°C. Stainless Steel Sloping Lid avoids contamination, reduces evaporation and saves energy.
Brand: Grant Instruments™ LU28
2096.53 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- For use with Sub Aqua Pro Series Unstirred Water Bath
- Triangular-shaped stainless steel with plastic handle
- Choose from the following available sizes:
- 5L (LU6)
- 12L (LU14)
- 18 or 26L (LU28)
- 34L (LU36)
Specifications
SAP18, ST12, ST26 | |
Stainless Steel |
Product Content Correction
Product Title
Grant Instruments™ Stainless Steel Sloping Lids > For Use With: SAP18
Spot an opportunity for improvement?Share a Content Correction