Learn More
Buerkle™ Stainless Steel Spatula
Universal instrument for collecting, dividing and mixing pastes, creams etc.
158.00 DKK - 255.00 DKK
Specifications
Product Type | Spatula |
---|---|
For Use With (Application) | Universal Instrument for Collecting, Dividing and Mixing Pastes, Creams |
Blade Material | Stainless Steel V2A |
Handle Material | Wood |
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | ||||||
11878502
|
Buerkle™
5313-0180 |
180 mm |
158.00 DKK
Each |
Estimated Shipment: 20-08-2025
Log in to see stock. |
||||||
11971551
|
Buerkle™
5313-0300 |
300 mm |
255.00 DKK
Each |
Estimated Shipment: 06-08-2025
Log in to see stock. |
||||||
Description
Universal instrument for collecting, dividing and mixing pastes, creams etc.- Blade made of stainless steel V2A (1.4301)
- Wooden handle
Specifications
Spatula | |
Universal Instrument for Collecting, Dividing and Mixing Pastes, Creams | |
Stainless Steel V2A | |
Wood |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Buerkle™ Stainless Steel Spatula