Learn More
Stainless Steel Spheres, 2mm (0.08 in ) dia., Thermo Scientific Chemicals
212.85 DKK - 533.88 DKK
Specifications
Chemical Name or Material | Stainless Steel |
---|---|
Appearance | Spheres |
Molecular Formula | Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt% |
MDL Number | MFCD00148499 |
Form | Sphere |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11305959
|
Thermo Scientific Alfa Aesar
042019.KM |
25 Pc. | N/A | ||||||
11315959
|
Thermo Scientific Alfa Aesar
042019.KN |
100 Pc. |
533.88 DKK
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Stainless Steel | |
Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt% | |
Sphere |
Spheres | |
MFCD00148499 |
Safety and Handling
H317-H351: May cause an allergic skin reaction. Suspected of causing cancer.
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures