Learn More
Karl Hecht™ Stainless Steel Square Wire Baskets
An excellent option for general storage applications in the laboratory.
902.00 DKK - 1120.00 DKK
Specifications
Product Type | Wire Basket |
---|---|
Description | Wire mesh construction |
Material | Stainless Steel |
Shape | Square |
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
10328731
|
Karl Hecht™
41075016 |
16 x 16 x 16 cm |
902.00 DKK
Each |
Estimated Shipment: 20-08-2025
Log in to see stock. |
||||||
10669431
|
Karl Hecht™
41075020 |
20 x 20 x 20 cm |
1120.00 DKK
Each |
Estimated Shipment: 12-08-2025
Log in to see stock. |
||||||
Description
- Wire mesh construction so contents are visible
- Square shape
- Available in a range of sizes
Specifications
Wire Basket | |
Wire mesh construction | |
Stainless Steel | |
Square |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Karl Hecht™ Stainless Steel Square Wire Baskets