Learn More
asecos™ Stainless Steel Storage Can with Screw Cap
The Stainless Steel Safety Canister is ideal for the safe distribution and dosing of flammable liquids.
Brand:  asecos™ 40082
 View more versions of this product
Product Code. 17856346
  • 1145.00 DKK / Each
Estimated Shipment: 28-01-2025
to see stock.
Explore more special offers
Description
  • With screw cap (secured against loss) overpressure valve, PTFE seal, integrated flame barrier (removable) and plastic protective ring
  • Incl. multilingual safety sticker to designate the contents
Specifications
Specifications
0.5 gal.
Gray
6.22 in.
8.90 in.
2.01 lb.
2 L
Stainless Steel
158.00 mm
226.00 mm
0.91 kg