Learn More
Socorex™ Stainless Steel Straight Drenchers
Use Socorex™ Stainless Steel Straight Drenchers for animal feeding and dosing. These precision-assembled stainless-steel feeding needles are ideal for the delivery of fluid and are reusable since they are autoclavable.
Brand: Socorex™ 376.16102S
This item is not returnable.
View return policy
Description
- Stainless Steel
- Various lengths to accommodate feeding needs
- Autoclavable
- Reusable
Specifications
Autoclavable | |
Drenching Cannula | |
Stainless Steel | |
102 mm |
Product Content Correction
Product Title
Socorex™ Stainless Steel Straight Drenchers > Length: 102mm
Spot an opportunity for improvement?Share a Content Correction