Learn More
Usbeck Laborgeraete™ Stainless Steel Support Rods
Brand:  Usbeck Laborgeraete™ 2150
 View more versions of this product
Product Code. 10239101
  • 71.60 DKK / Each
Estimated Shipment: 06-05-2025
to see stock.
Explore more special offers
Specifications
Stainless Steel (18/10)
12 mm
Without Thread
450 mm
Support Rod
1 Ea.
Product Title
Usbeck Laborgeraete™ Stainless Steel Support Rods > Length: 450mm; without Thread