Learn More
Usbeck Laborgeraete™ Stainless Steel Support Rods
Brand:  Usbeck Laborgeraete™ 2154
 View more versions of this product
Product Code. 10629522
  • 148.42 DKK / Each
Only null left
Estimated Shipment: 23-07-2025
Specifications
Stainless Steel (18/10)
12 mm
Without Thread
1000 mm
Support Rod
1 Ea.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Usbeck Laborgeraete™ Stainless Steel Support Rods > Length: 1000mm; without Thread