Learn More
VACUUBRAND™ Stainless Steel T-Pieces With Small KF Flanges
920.00 DKK - 1955.00 DKK
Specifications
For Use With Solenoid-operated valves VV and VV C
Material Stainless Steel
Quantity 1 Each
Type T-piece
View More Specs
Products 4
Product Code Brand Diameter (Metric) Inside Price Quantity & Availability  
Product Code Brand Diameter (Metric) Inside Price Quantity & Availability  
10450463
View Documents
VACUUBRAND™
20673514
16/16/16 mm
1075.00 DKK
Each
Only null left
Estimated Shipment: 30-06-2025
Add to basket
10059570
View Documents
VACUUBRAND™
20673500
10/10/10 mm
920.00 DKK
Each
Only null left
Estimated Shipment: 01-07-2025
Add to basket
10009600
View Documents
VACUUBRAND™
20673515
25/25/25 mm
1370.00 DKK
Each
Only null left
Estimated Shipment: 02-07-2025
Add to basket
10682222
View Documents
VACUUBRAND™
20673516
40/40/40 mm
1955.00 DKK
Each
Only null left
Estimated Shipment: 30-07-2025
Add to basket
Description
Description
Specifications
Specifications
Solenoid-operated valves VV and VV C
Stainless Steel
1 Each
T-piece
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
VACUUBRAND™ Stainless Steel T-Pieces With Small KF Flanges