Learn More
RETSCH Stainless-Steel Test Sieves, 200 Dia. x 50 mmH, in Micrometer Pore with ASTM E 11
Brand:  RETSCH 60.132.000250
 View more versions of this product
Product Code. 10228691
  • 938.00 DKK / Each
Only null left
Estimated Shipment: 18-09-2025
Specifications
1 Each
Test Sieve
Shaker AS 200
50mm
200 mm
Stainless Steel
250 μm
ASTM E 11
Certificate indicating Compliance
For separation, fractioning, particle size determination
Stainless Steel
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
RETSCH Stainless-Steel Test Sieves, 200 Dia. x 50 mmH, in Micrometer Pore with ASTM E 11 > Pore Size: 250 μm