Learn More
Fisherbrand™ Stainless Steel Tripods
Obtain optimum results with this stable and portable stainless steel tripod. Strong and light weight, this FisherbrandTM Stainless Steel Tripod features an adjustable height that allows it to be used by most people.
Brand:  Fisherbrand™ TR3
 View more versions of this product
Product Code. 10448402
  • 72.50 DKK / Each
Only null left
Estimated Shipment: 04-09-2025
Add to basket
Description
Description
  • Stable and portable platform for mount
  • Adjustable height allows to be used by most people
  • Easy to transport
  • Strong and light weight
  • Used as portable mount in a variety of applications
Specifications
Specifications
Stainless Steel
150 mm
Tripod
1 Ea.
210 mm
Plastic Feet
Bright
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Fisherbrand™ Stainless Steel Tripods > Height: 210mm; Width: 150mm