Learn More
Fisherbrand™ Scaffolding Tubes with End Plugs
Provides for sturdy scaffolding construction in the laboratory. Fisherbrand™ Scaffolding Tubes with End Plugs are manufactured from durable stainless steel.
Brand:  Fisherbrand™ 2121/33
 View more versions of this product
Product Code. 11582662
  • 107.00 DKK / Each
Only null left
Estimated Shipment: 13-10-2025
Add to basket
Description
Description
  • Approx. 13mm diameter
  • Available in a range of lengths
  • Fits with Fisherbrand scaffolding connectors and foot adapters
Specifications
Specifications
Stainless Steel
250 mm
Tube
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Fisherbrand™ Scaffolding Tubes with End Plugs > Length: 250mm