Learn More
Corning™ Standard Conical Bioprinting Nozzles
These Standard Conical Bioprinting Nozzles are designed fo use with Matribot™ Bioprinter. They are made from polypropylene, have a female Luer connector, and are available in 22G, 25G, and 27G.
Brand: Corning™ 6165
This item is not returnable.
View return policy
Description
- Made of polypropylene
- Feature a female Luer connector
- Available in 22 G, 25 G, and 27 G.
Specifications
Clear | |
Polypropylene | |
Feature Female Luer Connector | |
1.26 in. | |
50 Nozzles |
Bioprinting Nozzle | |
200 μm | |
27 G | |
32 mm |
Product Suggestions
Customers who viewed this item also viewed.
Product Content Correction
Product Title
Corning™ Standard Conical Bioprinting Nozzles > Conical Bioprinting Nozzle; Gauge: 27 G; Nozzle Diameter (Metric): 200 μm; Color: Clear
Spot an opportunity for improvement?Share a Content Correction