Learn More
SCHOTT AG Lighting and Imaging™ Standard controller MC 1000 for VisiLED Series
The VisiLED Controllers are the core of the VisiLED system to control the LED light heads
Brand: SCHOTT AG Lighting and Imaging™ 400080
This item is not returnable.
View return policy
Description
Standard controller MC 1000 allows settings of various illumination parameters including light intensity, 4 segment modes and change of illumination direction.
Specifications
Controller | |
Stereo Microscopy Illumination |