Learn More
Standard Precision Buffer Solution pH 9.225, Reagecon™
Brand: Reagecon 109225
Quantity | 500 mL |
---|
This item is not returnable.
View return policy
Specifications
500 mL | |
9.225 |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
1922524D1 | Certificate of Analysis | 22/01/2025 | 109225 |
1922523K1 | Certificate of Analysis | 09/04/2024 | 109225 |
1922523B1 | Certificate of Analysis | 14/02/2024 | 109225 |
1922522A1 | Certificate of Analysis | 14/09/2022 | 109225 |
1922521J2 | Certificate of Analysis | 29/03/2022 | 109225 |
Product Content Correction
Product Title
Standard Precision Buffer Solution pH 9.225, Reagecon™ > Quantity: 500mL
Spot an opportunity for improvement?Share a Content Correction