Learn More
Spectrum™ Labs Standard RC Dialysis Disc
For standard grade RC dialysis tubing
1925.00 DKK - 1926.78 DKK
Specifications
Product Type | Standard RC Dialysis Disc |
---|---|
Quantity | 50 Pk. |
For Use With (Application) | For standard grade RC dialysis tubing |
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | ||||||
12727526
|
Spectrum™ Labs
132482 |
100 mm |
1925.00 DKK
Pack of 50 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
12767526
|
Spectrum™ Labs
132484 |
33 mm | N/A | N/A | N/A | |||||
Description
For use with Spectrum Equilibrium Dialyzer and Spectra/Por™ MacroDialyzer systems.- Regenerated cellulose (RC) precut membrane discs
- Supplied dry with a glycerol as a humectant
Specifications
Standard RC Dialysis Disc | |
50 Pk. | |
For standard grade RC dialysis tubing |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Spectrum™ Labs Standard RC Dialysis Disc