Learn More
Macherey-Nagel™ Silica 60 Standard Adsorbent for Column Chromatography
Use silca 60 for standard separations. Silica 60 Standard Adsorbent for Column Chromatography comes in a variety of particle sizes and package sizes.
Brand:  Macherey-Nagel™ 815350.5
 View more versions of this product
Product Code. 10450522
  • 1660.00 DKK / 5kg
Estimated Shipment: 09-05-2025
to see stock.
Explore more special offers
Description
  • Pore size ∼ 60 Å
  • Pore volume ∼ 0.75 mL/g
  • Spec. surface BET ∼ 500 m2/g;
  • Highly porous
  • Amorphous silicic acid in the form of hard, opalescent particles
  • Prepared by precipitation of water glass with sulfuric acid
Specifications
Specifications
0.2 to 0.5 mm
Column Chromatography
5 kg
∼500 m2/g
Column Chromatography
∼60 Å
Plastic Container
Silica
Standard Silica 60 Adsorbent
Product Title
Macherey-Nagel™ Silica 60 Standard Adsorbent for Column Chromatography > Quantity: 5kg; Paritcle Size: 0.2 to 0.5mm