Learn More
Bellingham Stanley™ Stanley Metal Tube
Brand: Bellingham Stanley™ 35-11
3372.35 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
Polarimeter Tube | |
Metal | |
Polarimeter | |
200 mm |
Product Content Correction
Product Title
Bellingham Stanley™ Stanley Metal Tube > For use with: For Polarimetric
Spot an opportunity for improvement?Share a Content Correction