Learn More
Fisherbrand™ Cut Out Rubber Covered Rings
With cut out section to allow easy removal of equipment.
Brand:  Fisherbrand™ 2015/11
 View more versions of this product
Product Code. 11745626
  • 198.00 DKK / Each
Only null left
Estimated Shipment: 26-09-2025
Add to basket
Description
Description
  • With rubber covered rings
  • Stem length to the center of bosshead is approximately 60mm
  • Constructed in mild steel with welded joints
  • Zinc-plated
Specifications
Specifications
Mild Steel
Stativring
Zinc Plated, Rubber Coated
100 mm
Cut out section
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Fisherbrand™ Cut Out Rubber Covered Rings > Outer Diameter: 100mm